KEY465198085 Key Organics / BIONET

N'-[(1Z)-(3-methylphenyl)methylidene]-3-(1H-pyrrol-1-yl)thiophene-2-carbohydrazide

  • MDL number MFCD02102723
  • Cas Number : 478063-23-9
  • Molecular Weight : 309.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83447

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-415S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart