KEY465198087 Key Organics / BIONET

N'-[(1Z)-(2,4-dichlorophenyl)methylidene]-2-(1H-pyrrol-1-yl)benzohydrazide

  • MDL number MFCD01873892
  • Cas Number : 344934-51-6
  • Molecular Weight : 358.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83449

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-419S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart