KEY465198088 Key Organics / BIONET

2,6-di-tert-butyl-4-[5-(methylsulfanyl)-1,3,4-thiadiazol-2-yl]phenol

  • MDL number MFCD01871581
  • Cas Number : 130116-26-6
  • Molecular Weight : 336.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83454

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6P-511S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart