KEY465198092 Key Organics / BIONET

4-chloro-N-{2-[(4-methyl-1,2,3-thiadiazol-5-yl)sulfanyl]ethyl}benzene-1-sulfonamide

  • MDL number MFCD02186708
  • Cas Number : 478063-53-5
  • Molecular Weight : 349.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83483

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-582S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart