KEY465198093 Key Organics / BIONET

methyl 1-(3-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylate

  • MDL number MFCD01871606
  • Cas Number : 478063-66-0
  • Molecular Weight : 264.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83501

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-674S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart