KEY465198100 Key Organics / BIONET

1-(4-chlorophenyl)-3-(4H-1,2,4-triazol-4-yl)thiourea

  • MDL number MFCD02187194
  • Cas Number : 5102-35-2
  • Molecular Weight : 253.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83540

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart