KEY465198112 Key Organics / BIONET

ethyl 1-[(2E)-2-cyano-2-[(dimethylamino)methylidene]acetyl]-3-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD02187328
  • Cas Number : 477713-80-7
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83568

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart