KEY465198117 Key Organics / BIONET

(2Z)-3-phenyl-2-[(4H-1,2,4-triazol-4-yl)imino]-1,3-thiazolidin-4-one

  • MDL number MFCD02570803
  • Cas Number : 5181-23-7
  • Molecular Weight : 259.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83577

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0061_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart