KEY465198125 Key Organics / BIONET

(2E)-1-[5-(2,4-dichlorophenyl)furan-2-yl]-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD02187209
  • Cas Number : 478064-05-0
  • Molecular Weight : 310.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83598

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0087_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart