KEY465198131 Key Organics / BIONET

N-[(3-benzylquinoxalin-2-yl)amino]-2-cyanoethanimidamide

  • MDL number MFCD02187218
  • Cas Number : 478064-19-6
  • Molecular Weight : 316.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83621

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6R-0213_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart