KEY465198132 Key Organics / BIONET

4-chloro-N'-{1H,2H-naphtho[2,1-b]furan-2-carbonyl}benzohydrazide

  • MDL number MFCD02187221
  • Cas Number : 478064-22-1
  • Molecular Weight : 366.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83625

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0219_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart