KEY465198144 Key Organics / BIONET

5-acetyl-4-(4-bromophenyl)-6-methyl-1,2,3,4-tetrahydropyridin-2-one

  • MDL number MFCD02661885
  • Cas Number : 252875-50-6
  • Molecular Weight : 308.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83680

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6R-0295_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart