KEY465198151 Key Organics / BIONET

N-[(cyclohexylcarbamoyl)amino]-3-(pyridin-3-yl)-4,5-dihydro-1,2-oxazole-5-carboxamide

  • MDL number MFCD02570966
  • Cas Number : 478064-65-2
  • Molecular Weight : 331.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83727

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6R-0356_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart