KEY465198158 Key Organics / BIONET

2-ethoxy-6-(furan-2-yl)pyridine-3-carbonitrile

  • MDL number MFCD02570837
  • Cas Number : 190579-90-9
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83748

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6R-0621_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart