KEY465198163 Key Organics / BIONET

4-[2-(4-chlorophenyl)-1,3-thiazol-5-yl]-N-(4-methyl-1,3-thiazol-2-yl)pyrimidin-2-amine

  • MDL number MFCD28347942
  • Cas Number : 1823183-28-3
  • Molecular Weight : 385.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 290 - 292
Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453090

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0804_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart