KEY465198189 Key Organics / BIONET

4-chloro-N-[2-(1H-indol-3-yl)ethyl]benzene-1-sulfonamide

  • MDL number MFCD01045667
  • Cas Number : 50626-41-0
  • Molecular Weight : 334.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83816

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-1028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart