KEY465198200 Key Organics / BIONET

(2E)-1-(4-chlorophenyl)-3-[2-(methylsulfanyl)quinolin-3-yl]prop-2-en-1-one

  • MDL number MFCD04125826
  • Cas Number : 866049-59-4
  • Molecular Weight : 339.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83838

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-1134_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart