KEY465198201 Key Organics / BIONET

(2E)-1-(4-fluorophenyl)-3-[2-(methylsulfanyl)quinolin-3-yl]prop-2-en-1-one

  • MDL number MFCD02571590
  • Cas Number : 478065-16-6
  • Molecular Weight : 323.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83839

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6R-1136_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart