KEY465198209 Key Organics / BIONET

1-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]ethyl 4-chlorobenzoate

  • MDL number MFCD04125833
  • Cas Number : 866049-64-1
  • Molecular Weight : 392.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83876

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-1210_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart