KEY465198210 Key Organics / BIONET

2-({4-ethyl-5-[(E)-2-phenylethenyl]-4H-1,2,4-triazol-3-yl}sulfanyl)acetonitrile

  • MDL number MFCD02571226
  • Cas Number : 478065-57-5
  • Molecular Weight : 270.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83878

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-1252_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart