KEY465198219 Key Organics / BIONET

2-{[4-(thiophen-2-yl)pyrimidin-2-yl]sulfanyl}acetonitrile

  • MDL number MFCD03102526
  • Cas Number : 692287-21-1
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83927

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart