KEY465198231 Key Organics / BIONET

2-({1-[(2-chloro-1,3-thiazol-5-yl)methyl]-1H-indol-3-yl}methylidene)propanedinitrile

  • MDL number MFCD03617672
  • Cas Number : 866049-74-3
  • Molecular Weight : 324.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83963

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6T-0077_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart