KEY465198264 Key Organics / BIONET

2-[(E)-[(4-chlorophenyl)methylidene]amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile

  • MDL number MFCD00808825
  • Cas Number : 1346414-21-8
  • Molecular Weight : 300.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84063

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6T-0631_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart