KEY465198265 Key Organics / BIONET

5-(methoxymethyl)-2-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-4-yl]pyrazolo[1,5-a]pyrimidin-7-ol

  • MDL number MFCD03617692
  • Cas Number : 1007074-04-5
  • Molecular Weight : 327.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84072

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6T-0807_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart