KEY465198321 Key Organics / BIONET

[(E)-[(3-methoxy-4-{[3-(trifluoromethyl)phenyl]methoxy}phenyl)methylidene]amino]thiourea

  • MDL number MFCD03787585
  • Cas Number : 866050-63-7
  • Molecular Weight : 383.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84247

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6W-0844_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart