KEY465198335 Key Organics / BIONET

8-(2-methoxyphenyl)-5,12,14-trioxa-2-azatetracyclo[7.7.0.0^{3,7}.0^{11,15}]hexadeca-1(9),3(7),10,15-tetraen-6-one

  • MDL number MFCD06496186
  • Cas Number : 866050-82-0
  • Molecular Weight : 337.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84276

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6X-0051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart