KEY465198343 Key Organics / BIONET

(2Z)-2-[(3-nitrophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one

  • MDL number MFCD02989619
  • Cas Number : 875299-80-2
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84299

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6X-0240_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart