KEY465198369 Key Organics / BIONET

2-(4-bromophenyl)-8-nitroimidazo[1,2-a]pyridine

  • MDL number MFCD06496189
  • Cas Number : 866051-28-7
  • Molecular Weight : 318.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84405

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6X-0882_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart