KEY465198450 Key Organics / BIONET

5-(4-fluorobenzoyl)-N-phenyl-1,3-thiazol-2-amine

  • MDL number MFCD00139533
  • Cas Number : 343376-29-4
  • Molecular Weight : 298.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7D-098_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart