KEY465198464 Key Organics / BIONET

N-[(3-chlorophenyl)methyl]-5-nitroimidazo[2,1-b][1,3]thiazol-6-amine

  • MDL number MFCD00138983
  • Cas Number : 339008-17-2
  • Molecular Weight : 308.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84735

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7D-129_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart