KEY465198553 Key Organics / BIONET

5-{1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]ethyl}-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00244380
  • Cas Number : 339010-57-0
  • Molecular Weight : 384.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85107

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7G-033_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart