KEY465198562 Key Organics / BIONET

4-methoxy-N-(propan-2-yl)-6-(trifluoromethyl)pyrimidin-2-amine

  • MDL number MFCD00172057
  • Cas Number : 339011-19-7
  • Molecular Weight : 235.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85182

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7G-359S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart