KEY465198600 Key Organics / BIONET

2-amino-4-(4-fluorophenyl)-6-phenylpyridine-3-carbonitrile

  • MDL number MFCD00244193
  • Cas Number : 126202-91-3
  • Molecular Weight : 289.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85359

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7H-301S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart