KEY465198671 Key Organics / BIONET

methyl 2-(3-chlorophenyl)-4-(4-chlorophenyl)-4-oxobutanoate

  • MDL number MFCD00664384
  • Cas Number : 344280-28-0
  • Molecular Weight : 337.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7J-528S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart