KEY465198701 Key Organics / BIONET

5-(3-chlorophenyl)-2-(prop-2-yn-1-ylsulfanyl)pyrimidine

  • MDL number MFCD00232556
  • Cas Number : 344281-98-7
  • Molecular Weight : 260.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85771

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7J-608S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart