KEY465198732 Key Organics / BIONET

2,4-dimethyl 3-(phenylcarbamoyl)-1,3-thiazolidine-2,4-dicarboxylate

  • MDL number MFCD00664226
  • Cas Number : 294848-66-1
  • Molecular Weight : 324.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85830

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7J-917_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart