KEY465198824 Key Organics / BIONET

6-chloro-N4,N4-diethylpyrimidine-2,4-diamine

  • MDL number MFCD00664696
  • Cas Number : 116895-09-1
  • Molecular Weight : 200.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-N~4~,N~4~-diethyl-2,4-pyrimidinediamine
Inchi Code 1S/C8H13ClN4/c1-3-13(4-2)7-5-6(9)11-8(10)12-7/h5H,3-4H2,1-2H3,(H2,10,11,12)
Inchi Key JZPXSIGRVGTURL-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86221

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7L-834_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart