KEY465198872 Key Organics / BIONET

2-(4-chlorobenzenesulfonamido)ethyl N-(4-chlorophenyl)carbamate

  • MDL number MFCD01315122
  • Cas Number : 478066-88-5
  • Molecular Weight : 389.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86410

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7M-553S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart