KEY465198897 Key Organics / BIONET

5-(4-fluorobenzoyl)-3-methylthieno[2,3-c][1,2]thiazol-4-amine

  • MDL number MFCD00974844
  • Cas Number : 339017-36-6
  • Molecular Weight : 292.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86498

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7M-733_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart