KEY465198983 Key Organics / BIONET

2-{[(2-chlorophenyl)methyl]sulfanyl}-N-methyl-5,6,7,8-tetrahydroquinazolin-4-amine

  • MDL number MFCD01568404
  • Cas Number : 339019-32-8
  • Molecular Weight : 319.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86808

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7N-467S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart