KEY465199018 Key Organics / BIONET

methyl 2-(3,4,4-trifluorobut-3-ene-1-sulfonyl)acetate

  • MDL number MFCD01443882
  • Cas Number : 339020-20-1
  • Molecular Weight : 246.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl [(3,4,4-trifluoro-3-butenyl)sulfonyl]acetate
Inchi Code 1S/C7H9F3O4S/c1-14-6(11)4-15(12,13)3-2-5(8)7(9)10/h2-4H2,1H3
Inchi Key LFCITJGUGBWWTG-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86949

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7N-758_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart