KEY465199068 Key Organics / BIONET

2-[4-(benzenesulfonyl)piperazin-1-yl]-1,3-benzothiazole

  • MDL number MFCD01626225
  • Cas Number : 309268-43-7
  • Molecular Weight : 359.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87125

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7P-361S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart