KEY465199100 Key Organics / BIONET

(2E)-3-[2,5-dimethyl-1-(4H-1,2,4-triazol-4-yl)-1H-pyrrol-3-yl]prop-2-enoic acid

  • MDL number MFCD02570900
  • Cas Number : 478077-83-7
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87234

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart