KEY465199143 Key Organics / BIONET

N'-(benzenesulfonyl)-3-(pyridin-3-yl)-4,5-dihydro-1,2-oxazole-5-carbohydrazide

  • MDL number MFCD02570998
  • Cas Number : 478078-45-4
  • Molecular Weight : 346.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87320

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0248_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart