KEY465199155 Key Organics / BIONET

4-(4-fluorobenzoyl)-N-(2-methoxyethyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD02570939
  • Cas Number : 478078-61-4
  • Molecular Weight : 290.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87348

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0290_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart