KEY465199203 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}-3-(2-chlorophenyl)propanoic acid

  • MDL number MFCD02090709
  • Cas Number : 284493-66-9
  • Molecular Weight : 299.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(tert-butoxycarbonyl)-3-(2-chlorophenyl)-beta-alanine
Inchi Code 1S/C14H18ClNO4/c1-14(2,3)20-13(19)16-11(8-12(17)18)9-6-4-5-7-10(9)15/h4-7,11H,8H2,1-3H3,(H,16,19)(H,17,18)
Inchi Key JBKHFGREKMCWAU-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87462

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0655_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart