KEY465199215 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-[2-(pyridin-3-yl)-1,3-thiazol-5-yl]prop-2-en-1-one

  • MDL number MFCD28347958
  • Cas Number : 1823194-88-2
  • Molecular Weight : 259.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203 - 204
Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453108

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0826_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart