KEY465199223 Key Organics / BIONET

3-fluoro-4-(4-methanesulfonylpiperazin-1-yl)aniline

  • MDL number MFCD04125976
  • Cas Number : 864146-96-3
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87509

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-1030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart