KEY465199253 Key Organics / BIONET

1-(2-chloro-4-fluorophenoxy)-3-(4-methylbenzenesulfonyl)propan-2-ol

  • MDL number MFCD02571291
  • Cas Number : 478079-86-6
  • Molecular Weight : 358.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87597

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-1267_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart