KEY465199312 Key Organics / BIONET

N-ethyl-1-{[3-(trifluoromethyl)phenyl]methyl}-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD03305754
  • Cas Number : 866131-98-8
  • Molecular Weight : 319.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87763

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7T-0285_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart